Hexadecylamine for synthesis, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: 1-Hexadecylamine, n-Hexadecylamine, 1-Aminohexadecane

M-Clarity™ quality level = MQ200

8.22203.0100 8.22203.0500 8.22203.9050
8.22203.0100EA 40.2 GBP
8.22203.0100 8.22203.0500 8.22203.9050
Hexadecylamine for synthesis, Sigma-Aldrich®
Hexadecylamine
Formula: H₃C(CH₂)₁₄CH₂NH₂
MW: 241,46 g/mol
Boiling Pt: 330 °C (1013 hPa)
Melting Pt: 43…46 °C
Density: 0,7821 g/cm³ (60 °C)
Flash Pt: 140 °C
Storage Temperature: Ambient
MDL Number: MFCD00008158
CAS Number: 143-27-1
EINECS: 205-596-8
UN: 3259
ADR: 8,I

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 43 - 46 °C
Molar Mass 241.46 g/mol
Boiling Point 330 °C (1013 hPa)
Vapor Pressure 1.3 hPa (20 °C)
Flash Point 140 °C
Refractive Index 1.4373 (50 °C, 589 nm)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR