Hexadecylamine 90%

Supplier: Thermo Fisher Scientific
Danger

120510050 120511000 120515000 120510025
ACRO120510050EA 46 GBP
ACRO120510050 ACRO120511000 ACRO120515000 ACRO120510025
Hexadecylamine 90%
Hexadecylamine
Formula: H₃C(CH₂)₁₄CH₂NH₂
Boiling Pt: 330 °C (760 mmHg)
Melting Pt: 38…47 °C
Density: 0,7821 g/cm³ (60 °C)
Flash Pt: 140 °C
Storage Temperature: Ambient
MDL Number: MFCD00008158
CAS Number: 143-27-1
EINECS: 205-596-8
UN: 3259
ADR: 8,II

Order Now

Specification Test Results

Appearance White Low melting crystalline mass and/or chunks
Infrared spectrum Conforms
Melting point 38 to 47 °C
GC ≥88 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR