Dodecylamine 97%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
1-Dodecylamine, Dodecan-1-amine, 1-Aminododecane
A15515.22
A15515.36
A15515.A1
A15515.22EA
24.9
GBP
A15515.22
A15515.36
A15515.A1
Dodecylamine 97%
Dodecylamine
Purity: 97+%
CAS No.: 124-22-1
Molecular Formula: CH3(CH2)11NH2
Formula Weight: 185.36
Flash Point: 115°(239°F)
Melting Point: 27-31°
Boiling Point: 64-66°/0.05mm
Beilstein Reference: 1633576
UN No.: 3259
MDL No.: MFCD00008154
Formula:
H₃C(CH₂)₁₀CH₂NH₂ MW: 185,35 g/mol Boiling Pt: 64…66 °C (0,05 mmHg) Melting Pt: 27…31 °C Density: 0,802 Flash Pt: 115 °C (239 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008154 CAS Number: 124-22-1 EINECS: 204-690-6 UN: 3259 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...