Fluoroacetone 97%
Supplier: Thermo Scientific
|
Danger
|
151780010
151780050
ACRO151780010EA
135
GBP
ACRO151780010
ACRO151780050
Fluoroacetone 97%
Fluoroacetone
Formula:
C₃H₅FO MW: 76,07 g/mol Boiling Pt: 75 °C (760 mmHg) Density: 1,054 g/cm³ Flash Pt: 7 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00000451 CAS Number: 430-51-3 EINECS: 207-064-0 UN: 1992 ADR: 3,II |
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.3660 to 1.3700 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...