5-Chloro-2-pentanone 90%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
5-Chloropentan-2-one
388291000
388290250
ACRO388291000EA
234
GBP
ACRO388291000
ACRO388290250
5-Chloro-2-pentanone 90%
5-Chloro-2-pentanone
Formula:
C₅H₉ClO MW: 120,58 g/mol Boiling Pt: 172…175 °C (760 mmHg) Melting Pt: –48 °C Density: 1,053 g/cm³ Flash Pt: 35 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001008 CAS Number: 5891-21-4 EINECS: 227-565-8 UN: 1224 ADR: 3,III |
Specification Test Results
Appearance | Brown to black Liquid |
Infrared spectrum | Conforms |
GC | ≥88 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...