1-Chlorohexane 95%
Supplier: Thermo Fisher Scientific
|
Warning
|
404640050
404641000
404645000
ACRO404640050EA
37.2
GBP
ACRO404640050
ACRO404641000
ACRO404645000
1-Chlorohexane 95%
1-Chlorohexane
Formula:
C₆H₁₃Cl Boiling Pt: 135 °C Melting Pt: –94 °C Density: 0,87 g/cm³ Flash Pt: 26 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001018 CAS Number: 544-10-5 EINECS: 208-859-5 UN: 1993 ADR: 3,III Merck Index: 13,02163 |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4190 to 1.4210 (20°C, 589 nm) |
GC | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...