1,6-Dichlorohexane ≥98%
Supplier: Thermo Fisher Scientific
Warning
|
Purity: 98%
CAS No.: 2163-00-0
Molecular Formula: Cl(CH2)6Cl
Formula Weight: 155.07
Flash Point: 73°(163°F)
Refractive Index: 1.4568
Melting Point: -13°
Boiling Point: 204-205°
Beilstein Reference: 773673
UN No.: 3082
MDL No.: MFCD00001020
Formula:
ClH₂C(CH₂)₄CH₂Cl Boiling Pt: 204…205 °C Melting Pt: –13 °C Density: 1,067 Flash Pt: 73 °C (163 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001020 CAS Number: 2163-00-0 EINECS: 218-491-7 UN: 3082 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...