1,2-Dichloroethane, EMPLURA®, Supelco®

Supplier: Merck
Danger

Synonyms: Ethylene dichlorideDichloroethaneEthylene chloride

M-Clarity™ quality level = MQ200

1.00955.1000EA 0 GBP
1.00955.1000
1,2-Dichloroethane, EMPLURA®, Supelco®
1,2-Dichloroethane
Formula: ClH₂CCH₂Cl
MW: 98,96 g/mol
Boiling Pt: 83,5 °C (1013 hPa)
Melting Pt: –35,5 °C
Density: 1,254 g/cm³ (20 °C)
Flash Pt: 13 °C
Storage Temperature: Ambient
MDL Number: MFCD00000963
CAS Number: 107-06-2
EINECS: 203-458-1
UN: 1184
ADR: 3 (6.1),II
Merck Index: 13,03831
https://ni.vwr.com/stibo/web/std.lang.all/54/88/725488.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR