Acetonylacetone 97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Hexane-2,5-dione, 2,5-Hexanedione
102400050
102401000
102402500
102400010
ACRO102400050EA
31.3
GBP
ACRO102400050
ACRO102401000
ACRO102402500
ACRO102400010
Acetonylacetone 97%
Acetonylacetone
Formula:
C₆H₁₀O₂ MW: 114,14 g/mol Boiling Pt: 185…193 °C (760 mmHg) Melting Pt: –6 °C Density: 0,973 g/cm³ Flash Pt: 78 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008792 CAS Number: 110-13-4 EINECS: 203-738-3 Merck Index: 13,00072 |
Specification Test Results
Appearance | Clear yellow to brown Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4250 to 1.4270 (20°C, 589 nm) |
GC | ≥96.0 % |
BSC Certification | Passes |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...