Cryolite
Supplier: Merck
|
Danger
|
Synonyms:
Trisodium hexafluoroaluminate
1.06457.0100
1.06457.1000
1.06457.0100EA
0
GBP
1.06457.0100
1.06457.1000
Cryolite
Cryolite
Formula:
AlF₆Na₃ MW: 209,94 g/mol Melting Pt: 1000 °C Density: 2,9 g/cm³ (20 °C) Storage Temperature: Ambient |
CAS Number:
15096-52-3 EINECS: 239-148-8 UN: 3077 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...