Thiobenzamide ≥98%

Supplier: Thermo Scientific
Danger

Synonyms: Benzothioamide

A12973.14 A12973.22 A12973.36
A12973.14EA 33.8 GBP
A12973.14 A12973.22 A12973.36
Thiobenzamide ≥98%
Thiobenzamide

Purity: 98% CAS No.: 2227-79-4 Molecular Formula: C7H7NS Formula Weight: 137.21 Flash Point: 120°(248°F) Melting Point: 114-118° Beilstein Reference: 606021 UN No.: 2811 MDL No.: MFCD00008060

Formula: C₇H₇NS
MW: 137,21 g/mol
Boiling Pt: 245 °C (1013 hPa)
Melting Pt: 114…118 °C
Flash Pt: 120 °C (248 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008060
CAS Number: 2227-79-4
EINECS: 218-765-6
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR