2-Decanone for synthesis, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Decan-2-one
M-Clarity™ quality level = MQ200
8.18795.0025EA
85.3
GBP
8.18795.0025
8.18795.9999
2-Decanone for synthesis, Sigma-Aldrich®
2-Decanone
Formula:
C₁₀H₂₀O MW: 156,27 g/mol Boiling Pt: 211 °C (1013 hPa) Melting Pt: 3 °C Density: 0,82 g/cm³ (20 °C) Storage Temperature: Ambient |
CAS Number:
693-54-9 EINECS: 211-752-6 |
Specification Test Results
Solubility | (20 °C) insoluble |
Melting Point | 3 °C |
Molar Mass | 156.26 g/mol |
Boiling Point | 209 - 212 °C |
Flash Point | 86 °C |
Refractive Index | 1.4253 (20 °C, 589 nm) |
Density | 0.82 g/cm3 (20 °C) |
Synonyms | Methyl octyl ketone |
Hill Formula | C10H20O |
Chemical Formula | CH3(CH2)7COCH3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...