2-Decanone ≥97%
Supplier: Thermo Scientific
Warning
|
Synonyms:
Decan-2-one
A14145.14EA
62
GBP
A14145.14
A14145.06
2-Decanone ≥97%
2-Decanone
Formula:
C₁₀H₂₀O MW: 156,27 g/mol Boiling Pt: 210…212 °C Melting Pt: 3 °C Density: 0,824 Flash Pt: 86 °C (187 °F) Storage Temperature: Ambient |
CAS Number:
693-54-9 EINECS: 211-752-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...