Cyclopentyltriphenylphosphonium bromide ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
A14647.09
A14647.18
A14647.30
A14647.09EA
35.7
GBP
A14647.09
A14647.18
A14647.30
Cyclopentyltriphenylphosphonium bromide ≥98%
Cyclopentyltriphenylphosphonium bromide
Purity: 98%
CAS No.: 7333-52-0
Molecular Formula: C23H24BrP
Formula Weight: 411.33
Melting Point: 262-266°
Beilstein Reference: 3580193
Formula:
C₂₃H₂₄BrP Melting Pt: 262…266 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051880 CAS Number: 7333-52-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...