Carbazole

Supplier: Molekula
Warning

MOLE86934261-100GEA 138 GBP
MOLE86934261-100G
Carbazole
Carbazole
Formula: C₁₂H₉N
MW: 167,21 g/mol
Boiling Pt: 355 °C (1013 hPa)
Melting Pt: 236 °C
Density: 1,26 g/cm³ (20 °C)
MDL Number: MFCD00004960
CAS Number: 86-74-8
EINECS: 201-696-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR