5-Methyl-1-hexyne-3-ol ≥97%

Supplier: Thermo Scientific
Danger

Synonyms: 5-Methyl-1-hexyn-3-ol, 5-Methylhex-1-yn-3-ol, 3-Hydroxy-5-methyl-1-hexyne

H53490.03 H53490.06 H53490.14
H53490.03EA 32.6 GBP
H53490.03 H53490.06 H53490.14
5-Methyl-1-hexyne-3-ol ≥97%
5-Methyl-1-hexyne-3-ol
Formula: C₇H₁₂O
MW: 112,17 g/mol
Boiling Pt: 150…156 °C (1013 hPa)
Density: 0,860
Flash Pt: 53 °C (127 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00041606
CAS Number: 61996-79-0
UN: 1987
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR