2-Acetylfuran ≥99%

Supplier: Thermo Scientific
Danger

Synonyms: 2-Furyl methyl ketone, 1-(Furan-2-yl)ethanone

A14938.18 A14938.30
A14938.18EA 23.9 GBP
A14938.18 A14938.30
2-Acetylfuran ≥99%
2-Acetylfuran

Purity: 99% CAS No.: 1192-62-7 Molecular Formula: C6H6O2 Formula Weight: 110.11 Storage Temperature: 2°C to 8°C Flash Point: 71°(159°F) Refractive Index: 1.5070 Melting Point: 27-30° Boiling Point: 171-173° Beilstein Reference: 107909 UN No.: 2811 MDL No.: MFCD00003242

Formula: C₆H₆O₂
MW: 110,11 g/mol
Boiling Pt: 171…173 °C
Melting Pt: 27…30 °C
Density: 1,098
Flash Pt: 71 °C (159 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00003242
CAS Number: 1192-62-7
EINECS: 214-757-1
UN: 2811
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR