3-Chloro-5-fluoroaniline 97%
Supplier: Thermo Scientific
|
Warning
|
374740050
ACRO374740050EA
685
GBP
ACRO374740050
3-Chloro-5-fluoroaniline 97%
3-Chloro-5-fluoroaniline
Formula:
C₆H₅ClFN MW: 145,56 g/mol Density: 1,45 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD03407962 CAS Number: 4863-91-6 UN: 2810 ADR: 6.1,III |
Specification Test Results
Appearance | Clear light yellow to orange Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5610 to 1.5640 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...