1-Bromo-2-chloroethane ≥98%
Supplier: Thermo Fisher Scientific
|
Danger
|
A12349.0E
A12349.22
A12349.36
A12349.0EEA
465
GBP
A12349.0E
A12349.22
A12349.36
1-Bromo-2-chloroethane ≥98%
1-Bromo-2-chloroethane
Purity: 98%
CAS No.: 107-04-0
Molecular Formula: C2H4BrCl
Formula Weight: 143.42
Flash Point: None
Refractive Index: 1.4900
Odour: Sweet
Melting Point: -18°
Boiling Point: 106-107°
Beilstein Reference: 605265
UN No.: 2810
MDL No.: MFCD00000962
Formula:
BrH₂CCH₂Cl Boiling Pt: 106…107 °C Melting Pt: –18 °C Density: 1,726 Storage Temperature: Ambient |
MDL Number:
MFCD00000962 CAS Number: 107-04-0 EINECS: 203-456-0 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...