(±)-3-Methylcyclopentanone ≥99%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
DL-3-Methylcyclopentanone
L04940.03
L04940.06
L04940.03EA
26
GBP
L04940.03
L04940.06
(±)-3-Methylcyclopentanone ≥99%
(±)-3-Methylcyclopentanone
Purity: 99%
CAS No.: 1757-42-2
Molecular Formula: C6H10O
Formula Weight: 98.15
Flash Point: 36°(96°F)
Refractive Index: 1.4340
Melting Point: -58°
Boiling Point: 144-146°
Beilstein Reference: 1305074
UN No.: 1224
MDL No.: MFCD00001416
Formula:
C₆H₁₀O MW: 98.14 g/mol Boiling Pt: 144…146 °C Melting Pt: –58 °C Density: 0.914 Flash Pt: 36 °C (96 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001416 CAS Number: 1757-42-2 EINECS: 217-148-9 UN: 1224 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...