CAPS (3-(cyclohexylamino)propanesulphonic acid)

Supplier: Molekula

Synonyms: 3-Cyclohexylaminopropanesulphonic acid

24971012-500G 24971012-100G
MOLE24971012-500GEA 227 GBP
MOLE24971012-500G MOLE24971012-100G
CAPS (3-(cyclohexylamino)propanesulphonic acid)
CAPS (3-(Cyclohexylamino)propanesulphonic acid)
Formula: C₆H₁₁NH(CH₂)₃SO₃H
MW: 221,32 g/mol
Melting Pt: 324 °C
Storage Temperature: Ambient
MDL Number: MFCD00003837
CAS Number: 1135-40-6
EINECS: 214-492-1
Merck Index: 13,01773

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR