VWR®, Block Heaters, Digital

Supplier: VWR Collection
460-3264EA 956 GBP
460-3264 460-3266 460-3267 460-3269 460-3270 460-3272 460-3273 460-3275 460-3276 460-3278
VWR®, Block Heaters, Digital
Block Heaters Digital Block Heaters
Designed for applications that require repeatable results and superior temperature stability. These multipurpose units are ideal for incubation and activation of cultures, enzyme reactions, blood/urea nitrogen determinations, immunoassays, melting or boiling points. Easy to use controls with independent LED displays show actual and set point temperatures. Controls allow users to adjust temperature set point in ±0,1 °C increments. Timer function with audible alarm when time reaches zero. Optional external temperature probe kit monitors actual block or sample temperature. LED display on front panel will show actual probe temperature and adjust heater output to maintain user-defined set point. All dry block heaters include a built-in support rod holder for mounting the probe kit. Efficient heater features close tube-to-block contact with a low density design for exceptional temperature uniformity.

  • Exceptional temperature uniformity and stability. Units offer a temperature stability of ±0,2 °C
  • Temperature range from ambient 5 to 120 °C
  • Timer will display elapsed time or when programmed to user-defined limit, will count down to zero and shut off unit
  • Accommodate VWR® interchangeable modular heating blocks for tubes from 0,2 up to 50 ml capacity
  • Two LED displays, timer with audible alarm and microprocessor control

Ordering information: Heating blocks are not included, please order these separately.

Caution: * RT = Ambient
** EU-plug *** UK-plug **** CH-plug
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR