Tubing, silicone, Syndev®
Supplier: SF MEDICAL
SFM3-1050 SFM3-1550 SFM3-2650 SFM3-2950 SFM3-3650 SFM3-3350 SFM3-4850 SFM3-4950
228-0253EA 260 GBP
228-0253 228-0254 228-0256 228-0257 228-0259 228-0258 228-0261 228-0262
Tubing, silicone, Syndev®
Tubing Transfer Tubing
Silicone, translucent, medical grade, platinum cured.
  • Excellent operating range from −60 to +220 °C
  • Sterilisable by radiation, ethylene oxide, autoclavable
  • Manufactured to GMP in a clean room, FDA compliant to food grade regulation 177.2600 and certified to the biocompatibility requirements of USP Class VI
Suitable for medical applications where it is critical that no leachable by-products are present. Typical applications include peristaltic pump tubing, drug testing, bioanalysis, genetic processing, wound drains and many other non critical applications.
Temperature resistance: −54 to +204 °C for flexibility retention.
Hardness (Shore durometer A): 60 ±5