Haematocrit sealing compound
Supplier: Brand
749510
521-9105EA 44.1 GBP
521-9105
Haematocrit sealing compound
Seals Sealing Compounds
Non drying cement on a plastic tray.
  • For rapid sealing of several hundred micro haematocrit capillaries
  • Up to 24 capillaries can be placed vertically in numbered positions along the sides of the tray
NOTE: Hold the capillaries (about 2/3 full) near the empty end and stick the empty end into the cement with a slight twist to seal it.