Funnels for weighing cream or cheese
Supplier: JONES FR
MP711
611-0101EA 129 GBP
611-0101
Funnels for weighing cream or cheese
Funnels Cream Funnels
Borosilicate glass funnel and stopper with ground joint for cream weighing into butyrometers in accordance with BS 696.