Measuring cylinders, conical, soda lime glass
Supplier: JONES FR
CM364 CM368 CM371 CM373
213-0175EA 175 GBP
213-0175 213-0176 213-0177 213-0178
Measuring cylinders, conical, soda lime glass
Cylinders Measuring Cylinders
Soda-lime glass, clear.
  • Good chemical resistance
  • Broad base for stability
  • Liquid volume is easy to read
Ideal for a wide range of applications. With metric graduations. Broad base for stability.