2-Methyl-1-butene-3-yne 98%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
2-Methylbut-1-en-3-yne
336480100
336480500
ACRO336480100EA
177
GBP
ACRO336480100
ACRO336480500
2-Methyl-1-butene-3-yne 98%
2-Methyl-1-butene-3-yne
Formula:
C₅H₆ MW: 66,1 g/mol Boiling Pt: 32 °C Melting Pt: –113 °C Density: 0,69 g/cm³ Flash Pt: –45 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00008599 CAS Number: 78-80-8 EINECS: 201-144-9 UN: 3295 ADR: 3,I |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4140 to 1.4180 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...