3,5-Diamino-1H-1,2,4-triazole 98%
Supplier: Thermo Scientific
Synonyms:
3,5-Diamino-1,2,4-triazole, Guanazole
112440250
112441000
ACRO112440250EA
158
GBP
ACRO112440250
ACRO112441000
3,5-Diamino-1H-1,2,4-triazole 98%
3,5-Diamino-1H-1,2,4-triazole
Formula:
C₂H₅N₅ MW: 99,1 g/mol Boiling Pt: 425 °C (1013 hPa) Melting Pt: 204…208 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00005233 CAS Number: 1455-77-2 EINECS: 215-937-2 |
Specification Test Results
Appearance | White to beige to grey Crystalline powder or crystals and/or chunks |
Infrared spectrum | Conforms |
Melting point | 202 to 207 °C |
Titration with HClO4 | ≥97.5 % |
Calcium (Ca) | ≤200 ppm |
Iron (Fe) | ≤50 ppm |
Heavy metals (as Pb) | ≤50 ppm |
Insoluble matter | ≤0.01 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...