Sudan I, Supelco®
Supplier: Merck
Synonyms:
Solvent Yellow 14
, 1-(Phenyldiazenyl)naphthalen-2-ol
, Sudan Yellow
, Scarlet B
, Oil Orange
, 1-Phenylazo-2-naphthol
M-Clarity™ quality level = MQ100
51383-25MGEA
124
GBP
51383-25MG
Sudan I, Supelco®
Sudan I
Formula:
C₁₆H₁₂N₂O MW: 248,28 g/mol Boiling Pt: 444 °C (1013 hPa) Melting Pt: 135 °C Density: 1,119 g/cm³ (151 °C) |
MDL Number:
MFCD00003911 CAS Number: 842-07-9 EINECS: 212-668-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...