Mecamylamine hydrochloride 98%
Supplier: Thermo Fisher Scientific
|
Danger
|
470010010
470012500
470010010.EA
1200
GBP
470010010.
470012500.
Mecamylamine hydrochloride 98%
Mecamylamine hydrochloride
Formula:
C₁₁H₂₂ClN Storage Temperature: Refrigerator |
MDL Number:
MFCD00151462 CAS Number: 826-39-1 EINECS: 212-555-8 UN: 2811 ADR: 6.1,III |
Specification Test Results
Appearance | white to almost white powder |
Infrared spectrum | Conforms |
GC | ≥97.5 % |
Loss on drying | ≤1 % (105°C, 1 Hr, 5 mm Hg) |
Titration Argentometric | 17.0 to 17.8 % (Cl) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...