Perphenazine
Supplier: Thermo Fisher Scientific
|
Warning
|
468490010
468490050
468490250
468490010.EA
70.3
GBP
468490010.
468490050.
468490250.
Perphenazine
Perphenazine
Formula:
C₂₁H₂₆ClN₃OS Storage Temperature: Refrigerator |
MDL Number:
MFCD00056798 CAS Number: 58-39-9 EINECS: 200-381-5 |
Specification Test Results
Appearance | white to orange to green powder |
Infrared spectrum | Conforms |
Titration with HClO4 | 97.5 to 102.5 % |
Loss on drying | ≤0.5 % (1 g, 65°C, 4 h) (vacuum) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...