N,N-Diethylbenzylamine
Supplier: Thermo Fisher Scientific
|
Danger
|
468460250
468461000
468460250.EA
87.9
GBP
468460250.
468461000.
N,N-Diethylbenzylamine
N,N-Diethylbenzylamine
Formula:
C₁₁H₁₇N Density: 0,89 Flash Pt: 77°C Storage Temperature: Refrigerator |
MDL Number:
MFCD00026731 CAS Number: 772-54-3 EINECS: 212-251-5 UN: 2735 ADR: 8,III |
Specification Test Results
Appearance | clear colorless to light yellow liquid |
Infrared spectrum | Conforms |
GC | ≥97.5 % |
Refractive index | 1.4960 to 1.4990 (20°C, 589 nm) |
Titration with HClO4 | 97.5 to 102.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...