2-Methylindene

Supplier: Thermo Fisher Scientific

Synonyms: 2-Methyl-1H-indene

467760010 467760050
467760010.EA 107 GBP
467760010. 467760050.
2-Methylindene
2-Methylindene
Formula: C₁₀H₁₀
MW: 130.1900 g/mol
Density: 0.971
Flash Pt: 76°C
Storage Temperature: Ambient
MDL Number: MFCD00274253
CAS Number: 2177-47-1

Order Now

Specification Test Results

Appearance clear yellow liquid
Infrared spectrum Conforms
GC ≥97.5 %
Refractive index 1.5665 to 1.5675 (20°C, 589 nm)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR