Magnesium aluminate ≥99.9% (metals basis excluding Ca) (max. 25 ppm Ca)

Supplier: Thermo Scientific

Synonyms: Dialuminium magnesium tetraoxide, Magnesium aluminum oxide, Aluminum magnesium oxide

36388.14 36388.22
36388.14EA 0 GBP
36388.14 36388.22
Magnesium aluminate ≥99.9% (metals basis excluding Ca) (max. 25 ppm Ca)
Magnesium aluminate

Purity: 99.9% CAS No.: 12068-51-8 Molecular Formula: MgAl2O4 Formula Weight: 142.26 Physical Form: 3-12mm Fused Pieces Odour: Odourless MDL No.: MFCD00049475

Formula: Al₂MgO₄
MW: 142,27 g/mol
MDL Number: MFCD00049475
CAS Number: 12068-51-8
EINECS: 235-100-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR