Diisopropylcarbodiimide ≥99%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
N,N'-methanetetraylbis(1-methylethylamine), N,N-Diisopropylcarbodiimide, 1,3-Diisopropylcarbodiimide, N,N'-Diisopropylcarbodiimide, N.N’-Diisopropylcarbodiimide, DIC
A19292.09
A19292.14
A19292.18
A19292.30
A19292.09EA
39.2
GBP
A19292.09
A19292.14
A19292.18
A19292.30
Diisopropylcarbodiimide ≥99%
Diisopropylcarbodiimide
Purity: 99%
CAS No.: 693-13-0
Molecular Formula: (CH3)2CHN=C=NCH(CH3)2
Formula Weight: 126.20
Flash Point: 33°(91°F)
Refractive Index: 1.4330
Odour: Stench
Boiling Point: 145-148°
Beilstein Reference: 878281
UN No.: 2929
MDL No.: MFCD00065689
Formula:
C₇H₁₄N₂ MW: 126,2 g/mol Boiling Pt: 145…148 °C Density: 0,810 Flash Pt: 33 °C (91 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00065689 CAS Number: 693-13-0 EINECS: 211-743-7 UN: 2929 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...