4-(Methylsulphonyl)benzyl bromide 97
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
α-Bromo-4-(methylsulphonyl)toluene, 1-(Bromomethyl)-4-(methylsulphonyl)benzene
H50241.03
H50241.MD
H50241.03EA
0
GBP
H50241.03
H50241.MD
4-(Methylsulphonyl)benzyl bromide 97
4-(Methylsulphonyl)benzyl bromide
Purity: 97%
CAS No.: 53606-06-7
Molecular Formula: C8H9BrO2S
Formula Weight: 249.13
UN No.: 3261
MDL No.: MFCD00185849
Formula:
C₈H₉BrO₂S MW: 249.13 Melting Pt: 97 Storage Temperature: Ambient |
MDL Number:
MFCD00185849 CAS Number: 53606-06-7 UN: 3261 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...