Anti-Calretinin Rabbit Polyclonal Antibody

Supplier: ANTIBODIES.COM
A104325-100 A104325-50
ANTIA104325-100EA 474 GBP
ANTIA104325-100 ANTIA104325-50
Anti-Calretinin Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to Calretinin for WB, ICC/IF and IHC with samples derived from human, rat and mouse.

  • Validated Applications: WB, ICC/IF, IHC
  • Recommended Dilutions: WB: 1:5000 to 1:10000, ICC/IF: 1:5000 to 1:10000, IHC: 1:5000 to 1:10000

Type: Primary
Antigen: Calretinin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Rat;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR