Anti-MeCP2 Mouse Monoclonal Antibody [clone: 5H12]

Supplier: ANTIBODIES.COM
A104323-100 A104323-50
ANTIA104323-100EA 474 GBP
ANTIA104323-100 ANTIA104323-50
Anti-MeCP2 Mouse Monoclonal Antibody [clone: 5H12]
Antibodies
Mouse monoclonal (5A12) antibody to MeCP2 for WB, ICC/IF and IHC with samples derived from human, rat, mouse, canine, bovine, horse and monkey.

  • Validated Applications: WB, ICC/IF, IHC
  • Recommended Dilutions: WB: 1:5000 to 1:10000, ICC/IF: 1:1000 to 1:5000, IHC: 1:1000 to 1:5000

Type: Primary
Antigen: MeCP2
Clonality: Monoclonal
Clone: 5H12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human;Rat;Mouse;Dog;Bovine;Horse;Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR