Anti-Secretagogin Chicken Polyclonal Antibody

Supplier: ANTIBODIES.COM
A104317-100 A104317-50
ANTIA104317-100EA 474 GBP
ANTIA104317-100 ANTIA104317-50
Anti-Secretagogin Chicken Polyclonal Antibody
Antibodies
Chicken polyclonal antibody to Secretagogin for WB, ICC/IF and IHC with samples derived from human, rat and mouse.

  • Validated Applications: WB, ICC/IF, IHC
  • Recommended Dilutions: WB: 1:1,000 to 1:5,000, ICC/IF: 1:1,000 to 1:2,000, IHC: 1:1,000 to 1:2,000

Type: Primary
Antigen: Secretagogin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human, Rat, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR