Anti-PD-L1 Rabbit Recombinant Antibody [clone: DM124]

Supplier: ANTIBODIES.COM
A318583-100 A318583-500
ANTIA318583-100EA 583 GBP
ANTIA318583-100 ANTIA318583-500
Anti-PD-L1 Rabbit Recombinant Antibody [clone: DM124]
Antibodies
Recombinant rabbit monoclonal [DM124] antibody to PD-L1 for ELISA and flow cytometry with samples derived from human.

  • Validated applications: ELISA, Flow cytometry
  • Recommended dilutions: ELISA: 1:5000 to 10000, Flow cytometry: 1:100

Type: Primary
Antigen: PD-L1
Clonality: Recombinant
Clone: DM124
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR