Anti-AMHR2 Rabbit Recombinant Antibody [clone: DM186]

Supplier: ANTIBODIES.COM
A318523-100 A318523-500
ANTIA318523-100EA 583 GBP
ANTIA318523-100 ANTIA318523-500
Anti-AMHR2 Rabbit Recombinant Antibody [clone: DM186]
Antibodies
Recombinant rabbit monoclonal [DM186] antibody to AMHR2 for ELISA and Flow cytometry with samples derived from human.

  • Validated applications: ELISA, Flow cytometry
  • Recommended dilutions: ELISA: 1:5000 to 10000, Flow cytometry: 1:100

Type: Primary
Antigen: AMHR2
Clonality: Recombinant
Clone: DM186
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR