Anti-TRAP/CD40L Rabbit Recombinant Antibody [clone: DM164]

Supplier: ANTIBODIES.COM
A318545-100 A318545-500
ANTIA318545-100EA 583 GBP
ANTIA318545-100 ANTIA318545-500
Anti-TRAP/CD40L Rabbit Recombinant Antibody [clone: DM164]
Antibodies
Recombinant rabbit monoclonal [DM164] antibody to TRAP/CD40L for ELISA and Flow cytometry with samples derived from human.

  • Validated applications: ELISA, Flow cytometry
  • Recommended dilutions: ELISA: 1:5000 to 10000, Flow cytometry: 1:100

Type: Primary
Antigen: TRAP/CD40L
Clonality: Recombinant
Clone: DM164
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR