Ethylenetetracarbonitrile 98%
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
Tetracyanoethylene
138050100
138050250
ACRO138050100EA
165
GBP
ACRO138050100
ACRO138050250
Ethylenetetracarbonitrile 98%
Ethylenetetracarbonitrile
Formula:
C₆N₄ MW: 128.09 g/mol Boiling Pt: 223 °C (760 mmHg) Melting Pt: 196…201 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00001850 CAS Number: 670-54-2 EINECS: 211-578-0 UN: 3439 ADR: 6.1 I Merck Index: 12,09336 |
Specification Test Results
Appearance | White to beige to light brown Crystalline powder or crystals and/or chunks |
Infrared spectrum | Conforms |
Melting point | 196 to 201 °C |
Assay | ≥97.5 % (Ex N) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...