Anti-CD38 Mouse Monoclonal Antibody [clone: HIT2] (iFluor® 532)

Supplier: AAT BIOQUEST
10380070 10380071
AATB10380070EA 328 GBP
AATB10380070 AATB10380071
Anti-CD38 Mouse Monoclonal Antibody [clone: HIT2] (iFluor® 532)
Antibodies
HIT2 is an anti-human monoclonal antibody that targets the CD38 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Caution: For research use only.

Type: Primary
Antigen: CD38
Clonality: Monoclonal
Clone: HIT2
Conjugation: iFluor® 532
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR