Anti-CD3e Mouse Monoclonal Antibody [clone: SK7] (mFluor™ Green 620)

Supplier: AAT BIOQUEST
100330U0 100330U1
AATB100330U0EA 328 GBP
AATB100330U0 AATB100330U1
Anti-CD3e Mouse Monoclonal Antibody [clone: SK7] (mFluor™ Green 620)
Antibodies
The SK7 monoclonal antibody reacts with human CD3e, a 20 kD member of the Ig superfamily often found on the surface of t cells, tregs and nkt cells.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Caution: For research use only.

Type: Primary
Antigen: CD3e
Clonality: Monoclonal
Clone: SK7
Conjugation: mFluor™ Green 620
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR