Lauroyl chloride 98%
Supplier: Thermo Scientific
|
Danger
|
169030010
169032500
169030500
ACRO169030010EA
251
GBP
ACRO169030010
ACRO169032500
ACRO169030500
Lauroyl chloride 98%
Lauroyl chloride
Formula:
C₁₂H₂₃ClO Boiling Pt: 134…137 °C (11 mmHg) Melting Pt: –17 °C Density: 0,92 g/cm³ Flash Pt: 140 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000740 CAS Number: 112-16-3 EINECS: 203-941-7 UN: 3265 ADR: 8,III |
Specification Test Results
Appearance | Clear liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4440 to 1.4460 (20°C, 589 nm) |
GC | ≥97.5 % |
Color scale | ≤60 APHA |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...