Steel Ruler
Supplier: Radionics
RADS274-1070EA 95.8 GBP
RADS274-1070
Steel Ruler
Rulers
Steel rules for metric or imperial measurements.
  • Square end professional quality
  • Finish non-glare satin chrome
  • Manufactured from high quality steel
  • Fully hardened and tempered
Its sleek, satin finish exudes an air of refinement that immediately catches the eye. This measuring tool achieves the perfect balance of functionality and sophistication. Constructed from high-quality steel, it promises longevity, resisting scratches and wear with grace.
Rule markings:
Front side: 64ths, 32nds
Reverse side: 1,0 mm, 0,5 mm
Certifications: Rules conform to EEC-Class 1, Ref 73/362/EEC.