Anti-Alpaca IgG Goat Polyclonal Antibody (CF®680)

Supplier: Biotium

20887-100ULEA 265 GBP
20887-100UL
Anti-Alpaca IgG Goat Polyclonal Antibody (CF®680)
Antibodies
Goat anti-alpaca IgG antibody that recognizes the VHH domain of heavy chain alpaca IgG. Labeled with CF® Dyes and suitable for western or IF applications.

  • Available in 6 bright and photostable CF® Dyes
  • Suitable for western, immunofluorescence, and immunohistology in FFPE tissues
  • CF® Dye and biotin conjugates: add 0.5 mL distilled water
  • HRP or DNP conjugates: add 1 ml distilled water

Goat Anti-Alpaca IgG, VHH antibody is an affinity-purified polyclonal goat IgG whole antibody that recognizes the VHH domain of heavy chain alpaca IgG, subclasses 2 and 3, and the VHH domain of llama IgG, subclasses 2 and 3. The antibody is cross-adsorbed against bovine, human, mouse, rabbit, and rat serum proteins. It is not highly cross-adsorbed against immunoglobulins from other species. The antibody is available conjugated to Biotium’s bright and photostable CF® Dyes.

Type: Secondary
Antigen: Alpaca
Clonality: Polyclonal
Clone:
Conjugation: CF®680
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Alpaca, Llama
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR