Fumaronitrile 98%

Supplier: Thermo Fisher Scientific
Danger

157400050 157400250 157401000
ACRO157400050EA 102 GBP
ACRO157400050 ACRO157400250 ACRO157401000
Fumaronitrile 98%
Fumaronitrile
Formula: C₄H₂N₂
MW: 78.07 g/mol
Boiling Pt: 186 °C (760 mmHg)
Melting Pt: 95…98 °C
Storage Temperature: Ambient
MDL Number: MFCD00001928
CAS Number: 764-42-1
EINECS: 212-122-3
UN: 3439
ADR: 6.1 III

Order Now

Specification Test Results

Appearance White to cream Powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 95 to 99 °C
GC ≥97.5 %
Specific optical rotation +10° to +20° (20 °C, 589 nm) (c=4 in methanol)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR