Pyridine-2,4-dicarboxylic acid hydrate 99+%

Supplier: Thermo Scientific
Warning

Synonyms: Lutidinic acid hydrate, PCA hydrate, 2,4-Pyridinedicarboxylic acid hydrate

131860050 131860250
ACRO131860050EA 87.5 GBP
ACRO131860050 ACRO131860250
Pyridine-2,4-dicarboxylic acid hydrate 99+%
Pyridine-2,4-dicarboxylic acid
Formula: C₇H₅NO₄·xH₂O
MW: 167,12 g/mol (anhy)
Boiling Pt: 357 °C (1013 hPa)
Melting Pt: 243…246 °C (dec.)
Density: 0,942 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149413
CAS Number: 499-80-9
EINECS: 207-892-2

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Water ≤10 % (K.F.)
Titration with NaOH ≥99.0 % (On dry substance)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR